AlphaTec® MICROGARD 2000 Isolation Gowns
hvid, XL
Katalog # ANSEWH20B-00214-05
Leverandør: Ansell
Gå til:
Product Details & Documents
Superior breathable microporous laminate technology.
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Silicone-free
- Low-linting
The MICROGARD 2000 range is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0,01 µm in size. Bound seams offer superior strength, as well as a liquid and particle barrier.
These isolation gowns feature rear Velcro fastening, elasticated wrists, and bound seams.
Specifikationer
- Tøj størrelse:XL
- Farve:hvid
- Seq:04
- Cat. No.:ANSEWH20B-00214-05
- Sleeve or leg length:Long
- Model:Microgard 2000
- Outfit applikationer:Medical|Clinical



