Safety spectacles, i-works
Product Details & Documents
The 100% metal free uvex i-works encapsulates the best of uvex eyewear: the sporty design, panoramic lens and low weight make it extremely comfortable and together with the unrivalled permanent uvex lens coating technology (uvex supravision excellence), provides the highest level of optical clarity remaining fog free even after repeated cleaning. High-tech soft grip side arm materials further enhance comfort levels delivering a stable, non-slip fit.
- Sporty design and panoramic lens for great lateral vision
- High performance anti-fog coating on the inside, scratch resistant on the outside
- Comfortable stable, non-slip soft grip side arms
- Low profile side arms with practical hang cord attachment option
- 100% UV 400
Suitable for a range of applications requiring low impact protection including those working in construction, manufacturing, automotive, healthcare, utilities, pharmaceutical, food and beverages, warehouse and distribution.
Certifications: EN 166 (EN 172)
Documents
UVEX9194171 UVEX9194365
Images 360
Specifikationer
- Model:Anti-UV (5-2,5), anti-scratch, anti-fog
- Farve:koksgrå
- Seq:04
- Linser:Grey PC