Storage Bags
Food storage bag, 210×300 mm
Catalog # HKPK100128
Supplier: HK PACK KRAUTSCHEID
Jump to:
Product Details & Documents
LDPE transparent bags, ideal for storing and protecting food and small spare parts of all kinds.
- Food safety is ensured
- Flexible and tear resistant
- Freeze-resistant starting 50 μ and heat resistant up to 80 °C
Specifications
- Beskrivelse:Food storage bag
- Dimensioner:210×300 mm
- Seq:06