Orders LinkContactUsLinkComponent
Storage Bags
Storage Bags
Food storage bag, 300×500 mm
Katalog # HKPK100100
Leverandør:   HK PACK KRAUTSCHEID
Storage Bags
Katalog # HKPK100100
Leverandør:  HK PACK KRAUTSCHEID
Storage Bags
Storage Bags
Food storage bag, 300×500 mm
Price
1.770,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
LDPE transparent bags, ideal for storing and protecting food and small spare parts of all kinds.
  • Food safety is ensured
  • Flexible and tear resistant
  • Freeze-resistant starting 50 μ and heat resistant up to 80 °C
Specifikationer
  • Beskrivelse:
    Food storage bag
  • Dimensioner:
    300×500 mm
  • Seq:
    08