Orders LinkContactUsLinkComponent
Soxhlet extractors, Quickfit®
Soxhlet extractors, Quickfit®
Socket 75; cone 34/35; thimble size 60×160, 600 ml
Katalog # BIBBEX5/75
Leverandør:   SciLabware
Soxhlet extractors, Quickfit®
Katalog # BIBBEX5/75
Leverandør:  SciLabware
Soxhlet extractors, Quickfit®
Soxhlet extractors, Quickfit®
Socket 75; cone 34/35; thimble size 60×160, 600 ml
Price
9.820,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Borosilicate glass, large volume.
Flanged extractors which require Quickfit® or NS jointed flasks and condensers.
Specifikationer
  • Ekstraktionskap.:
    600 ml
  • Beskrivelse:
    Socket 75; cone 34/35; thimble size 60×160