Soxhlet extractors, Quickfit®
Socket 75; cone 34/35; thimble size 60×160, 600 ml
Katalog # BIBBEX5/75
Leverandør: SciLabware
Gå til:
Product Details & Documents
Borosilicate glass, large volume.
Flanged extractors which require Quickfit® or NS jointed flasks and condensers.
Specifikationer
- Ekstraktionskap.:600 ml
- Beskrivelse:Socket 75; cone 34/35; thimble size 60×160