Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
600 mm, 24/29 NS, 24/29 NS
Katalog # BIBBFC3/23
Leverandør:   SciLabware
Fractionating columns, Vigreux, Quickfit®
Katalog # BIBBFC3/23
Leverandør:  SciLabware
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
600 mm, 24/29 NS, 24/29 NS
Price
13.510,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
No glass jacket. With combination of horizontal and inclined indentations. Horizontal indentations ensure intimate vapour and piquid contatc. Inclined indentations redistribute liquid from walls to centre of column.
Specifikationer
  • Konusstr.:
    24/29 NS
  • Kappestr.:
    24/29 NS
  • Brugbar længde:
    600 mm
  • Seq:
    03