Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ChIP-Seq High Sensitivity Kit, Abcam

ChIP-Seq High Sensitivity Kit, Abcam

ChIP-Seq High Sensitivity Kit
Katalog # ABCAAB185908-12
Leverandør:   Abcam
ChIP-Seq High Sensitivity Kit, Abcam
Katalog # ABCAAB185908-12
Leverandør:  Abcam
CAS nummer:  
ChIP-Seq High Sensitivity Kit, Abcam

ChIP-Seq High Sensitivity Kit

CAS nummer:  
Price
9.640,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

ab185908 is a complete set of reagents required for carrying out a successful ChIP-Seq starting from mammalian cells or tissues.

Advarsel: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Beskrivelse:
    ChIP-Seq High Sensitivity Kit
  • Cat. No.:
    ABCAAB185908-12
  • Opbevaringstemperatur:
    Store at Multi.

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.