Orders LinkContactUsLinkComponent
Cylinders, borosilicate glass, with hexagonal base
Cylinders, borosilicate glass, with hexagonal base
250 ml
Katalog # SCOT213913679
Leverandør:   DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Katalog # SCOT213913679
Leverandør:  DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Cylinders, borosilicate glass, with hexagonal base
250 ml
Price
1.320,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
  • Without graduation
  • Hexagonal base
  • Base with three knobs which increase its stability
Certifications: DIN EN ISO 4788.
Specifikationer
  • Kapacitet:
    250 ml
  • Seq:
    06