Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(+)-Cellobiose ≥99% beta-glucosidase substrate

D-(+)-Cellobiose ≥99% beta-glucosidase substrate

Katalog # ABCAAB146391-5
Leverandør:   Abcam
D-(+)-Cellobiose ≥99% beta-glucosidase substrate
Katalog # ABCAAB146391-5
Leverandør:  Abcam
CAS nummer:  528-50-7
D-(+)-Cellobiose ≥99% beta-glucosidase substrate
CAS nummer:  528-50-7
Price
588,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Disaccharide consisting of two β-glucose molecules linked by a β-1,4 glycosidic bond. Used as a substrate of β-glucosidase.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymer:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS nummer:528-50-7
  • Molecular Weight:342,3 g/mol
Specifikationer

Specifikationstestresultater

  • Constituents::
    100% D-(+) Cellobiose
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.