Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Prodigiosin ≥95% apoptotic agent

Prodigiosin ≥95% apoptotic agent

Katalog # ABCAAB144331-1
Marketsource
Leverandør:   Abcam
Prodigiosin ≥95% apoptotic agent
Katalog # ABCAAB144331-1
Leverandør:  Abcam
Marketsource
CAS nummer:  
Prodigiosin ≥95% apoptotic agent
CAS nummer:  
Price
3.490,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents

Potent apoptotic agent. Antibiotic agent. Induces caspase-dependent apoptosis. Inhibits mTORC1 and mTORC2. Reactivates p53. Inhibits iNOS expression. Shows anti-ischemic and antimalarial effects in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • Molecular Weight:323,4 g/mol
Specifikationer

Specifikationstestresultater

  • Constituents::
    100% Prodigiosin