Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lanreotide

Lanreotide

Katalog # ABCAAB141209-5
Marketsource
Leverandør:   Abcam
Lanreotide
Katalog # ABCAAB141209-5
Leverandør:  Abcam
Marketsource
CAS nummer:  
CAS nummer:  
Price
823,00 kr
Log ind for tilgængelighed
1 * 5 mg
Product Details & Documents

MW 1096.3 Da, Purity >95%. Lanreotide, Somatostatin receptor analog. Achieve your results faster with highly validated, pure and trusted compounds.

  • Constituents: 100% [cyclo S-S]-3-(2- naphthyl)-D-alanyl-L-cysteinyl-L-tryrosyl-D-tryptophyl-L-lysyl-L-valyl-L- cysteinyl-L-threoninamide

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Cat. No.:
    ABCAAB141209-5
  • Protein/peptide name:
    Lanreotide
  • Molekylvægt:
    1096.3 DA
  • Renhed:
    >95%
  • Opbevaring betingelser:
    Store at -20 °C.
  • Shipping temperature:
    Ambient - Can Ship with Ice

Specifikationer