Lanreotide
Product Details & Documents
MW 1096.3 Da, Purity >95%. Lanreotide, Somatostatin receptor analog. Achieve your results faster with highly validated, pure and trusted compounds.
- Constituents: 100% [cyclo S-S]-3-(2- naphthyl)-D-alanyl-L-cysteinyl-L-tryrosyl-D-tryptophyl-L-lysyl-L-valyl-L- cysteinyl-L-threoninamide
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifikationer
Specifikationer
- Cat. No.:ABCAAB141209-1
- CAS-nr:1 mg
- Protein/peptide name:Lanreotide
- Molekylvægt:1096.3 DA
- Renhed:>95%
- Opbevaring betingelser:Store at -20 °C.
- Shipping temperature:Ambient - Can Ship with Ice



