Orders LinkContactUsLinkComponent
ECL Substrate Kit (High Sensitivity), Abcam
ECL Substrate Kit (High Sensitivity), Abcam
ECL Substrate Kit (High Sensitivity)
Catalog # ABCAAB133406-20
Supplier:   Abcam
ECL Substrate Kit (High Sensitivity), Abcam
Catalog # ABCAAB133406-20
Supplier:  Abcam
CAS Number:  
ECL Substrate Kit (High Sensitivity), Abcam
ECL Substrate Kit (High Sensitivity)
CAS Number:  
Price
784,00 kr
Log in for availability
1 * 20 mL
Product Details & Documents
High Sensitivity ECL Substrate Kit ab133406 is designed for the detection of proteins with 23pg-187ng of protein per band.
Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
  • Beskrivelse:
    ECL Substrate Kit (High Sensitivity)
  • Cat. No.:
    ABCAAB133406-20
  • Opbevaringstemperatur:
    Store at +4°C.
Specifications