Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

ECL Substrate Kit (High Sensitivity), Abcam

ECL Substrate Kit (High Sensitivity)
Katalog # ABCAAB133406-200
Marketsource
Leverandør:   Abcam
ECL Substrate Kit (High Sensitivity), Abcam
Katalog # ABCAAB133406-200
Leverandør:  Abcam
Marketsource
CAS nummer:  
ECL Substrate Kit (High Sensitivity), Abcam

ECL Substrate Kit (High Sensitivity)

CAS nummer:  
Price
2.590,00 kr
Log ind for tilgængelighed
1 * 200 mL
Product Details & Documents

High Sensitivity ECL Substrate Kit ab133406 is designed for the detection of proteins with 23pg-187ng of protein per band.

 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Beskrivelse:
    ECL Substrate Kit (High Sensitivity)
  • Cat. No.:
    ABCAAB133406-200
  • Opbevaringstemperatur:
    Store at +4°C.

Specifikationer