ESD insole, ESD-PRO
Insoles, ESD-Pro, full-length, 40, sort
114-7005
: ELTEN
Antibacterial and anatomically shaped ESD insole.
- Open cell structure of PU foam guarantees high moisture absorption
- Activated carbon absorber reduces odour
- Skin friendly materials improve the foot climate
For use with L10, Biomex (Renzo Biomex), Trainers, Sportics, Classic-Line, Men at Work, Safety-Grip, Business (Ronny), White. Suitable for ESD safety shoes (electrical resistance of 105 to 107 Ohm).
Colour: Black
- Skostørrelse:40
- Farve:sort
- Beskrivelse:Insoles, ESD-Pro, full-length
- Seq:06