ESD insole, ESD-PRO
Insoles, ESD-Pro, full-length, 37, sort
Katalog # 114-7002
Leverandør: ELTEN
Gå til:
Product Details & Documents
Antibacterial and anatomically shaped ESD insole.
- Open cell structure of PU foam guarantees high moisture absorption
- Activated carbon absorber reduces odour
- Skin friendly materials improve the foot climate
For use with L10, Biomex (Renzo Biomex), Trainers, Sportics, Classic-Line, Men at Work, Safety-Grip, Business (Ronny), White. Suitable for ESD safety shoes (electrical resistance of 105 to 107 Ohm).
Colour: Black
Specifikationer
- Skostørrelse:37
- Farve:sort
- Beskrivelse:Insoles, ESD-Pro, full-length
- Seq:03