Orders LinkContactUsLinkComponent
ESD insole, ESD-PRO
ESD insole, ESD-PRO
Insoles, ESD-Pro, full-length, 35, sort
Katalog # 114-7001
Leverandør:   ELTEN
ESD insole, ESD-PRO
Katalog # 114-7001
Leverandør:  ELTEN
ESD insole, ESD-PRO
ESD insole, ESD-PRO
Insoles, ESD-Pro, full-length, 35, sort
Price
36,60 kr
Log ind for tilgængelighed
1 * 1 Par
Product Details & Documents
Antibacterial and anatomically shaped ESD insole.
  • Open cell structure of PU foam guarantees high moisture absorption
  • Activated carbon absorber reduces odour
  • Skin friendly materials improve the foot climate
For use with L10, Biomex (Renzo Biomex), Trainers, Sportics, Classic-Line, Men at Work, Safety-Grip, Business (Ronny), White. Suitable for ESD safety shoes (electrical resistance of 105 to 107 Ohm).
Colour: Black
Specifikationer
  • Skostørrelse:
    35
  • Farve:
    sort
  • Beskrivelse:
    Insoles, ESD-Pro, full-length
  • Seq:
    01