Tannin
Glass bottle for solids, 1
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 1401-55-4 Molecular Formula: C76H52O46 Formula Weight: 1701.23 Odour: Odourless pH: 3.5 Melting Point: ca 218° dec. Beilstein Reference: 8186396 Merck Reference: 14,9052 MDL No.: MFCD00066397
Synonymer: Gallotannin, Tannin
- CAS nummer:1401-55-4
- Formel:C₇₆H₅₂O₄₆
- MDL nummer:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:14,9052
- Smeltende Pt:∼ 218 °C (dec.)
- Tæthed:>1 g/cm³ (20 °C)
- Flash Pt:199 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-753-2
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



