Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Industrial Scoops

Industrial Scoops

Industrial scoop, 200 ml, 225 mm, Rustfrit stål
Katalog # SAMPA643-200
Leverandør:   Sampling Systems
Industrial Scoops
Katalog # SAMPA643-200
Leverandør:  Sampling Systems
Industrial Scoops
Industrial Scoops

Industrial scoop, 200 ml, 225 mm, Rustfrit stål

Price
230,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Industrial scoops are cost-effective devices for general-purpose use.

  • Low-cost for general-purpose use
  • Made from stainless steel
  • Various sizes available
  • Available from stock
  • Fast delivery

Industrial scoops provide a low-cost solution for all types of general-purpose work. Available in different sizes, they are ideal for working with chemicals and most foodstuffs.

Specifikationer
  • Kapacitet:
    200 ml
  • Beskrivelse:
    Industrial scoop
  • Længde:
    225 mm
  • Materiale:
    Rustfrit stål
  • Seq:
    02