Combustion boats
103×21×14 mm
Katalog # TSLA75146
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Gå til:
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Specifikationer
- Seq:09
- B×D×H:103×21×14 mm