Orders LinkContactUsLinkComponent
Combustion boats
Combustion boats
103×21×14 mm
Katalog # TSLA75146
Combustion boats
Katalog # TSLA75146
Combustion boats
103×21×14 mm
Price
955,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
  • Very good chemical resistance
  • Very good temperature resistance
Specifikationer
  • Seq:
    09
  • B×D×H:
    103×21×14 mm