Orders LinkContactUsLinkComponent
Combustion boats
48×15×10 mm
  TSLA75143
Combustion boats
  TSLA75143
Combustion boats
48×15×10 mm
291,00 kr
1 * 1 Items
Combustion boats, quartz, glazed, with or without handle.
  • Very good chemical resistance
  • Very good temperature resistance
  • Seq:
    06
  • B×D×H:
    48×15×10 mm