Combustion boats
30×10×5 mm
Katalog # TSLA75141
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Gå til:
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Specifikationer
- Seq:04
- B×D×H:30×10×5 mm