ELECTRODE FILLING SOLUTION KCL 3M + AGCL 1 * 125 mL
Katalog # HACHLZW9511.99
Leverandør: Hach
Frequently Bought Together
Paraffin, Solidification point: 52-54°C, pellets, TECHNICAL
Leverandør: VWR Chemicals
VWR®, Nitril handsker, NITRILE
Leverandør: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
Vand USP, JP, Ph. Eur., purified
Leverandør: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Leverandør: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverandør: VWR Chemicals
Vinyl gloves, Semperguard®
Leverandør: SEMPERIT TECHN PROD