Glycin ≥99.5%
250 g
Electrophoresis Grade Heavy metals (as Pb): 20 ppm Used for preparation of Tris-glycine buffers.
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :Stue temperatur
- Pakn:250 g