Glycin ≥99.5%
Drum, 5 kg
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Electrophoresis Grade Heavy metals (as Pb): 20 ppm Used for preparation of Tris-glycine buffers.
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Storage Temperature:Stue temperatur
Specifications
Specifications
- Pakning:Drum
- Pakn:5 kg



