Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kolophonium, chunks

Kolophonium, chunks

1 kg
Katalog # ICNA0215655791
Leverandør:   MP Biomedicals
Kolophonium, chunks
Katalog # ICNA0215655791
Leverandør:  MP Biomedicals
CAS nummer:  8050-09-7
Kolophonium, chunks

1 kg

CAS nummer:  8050-09-7
Price
1.890,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Soluble in alcohol (ethanol: 50 mg/ml), benzene, ether. Insoluble in water. Chloroform: soluble 0,1 g/ml, clear, slightly yellow to deep yellow.

Soluble in alcohol (ethanol: 50 mg/ml), benzene, ether. Insoluble in water. Chloroform: soluble 0,1 g/ml, clear, slightly yellow to deep yellow.

Synonymer:  Terpentinoliefri harpiks
  • CAS nummer:8050-09-7
  • MDL nummer:MFCD00132205
  • Smeltende Pt:70…130 °C (lit.)
  • Smeltende Pt:70 - 130 °C (lit.)
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakn:
    1 kg

Specifikationer