Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

1,3-Propansulton, semi-solid

Glass bottle for solids, 25 g
Catalog # ICNA0215639125
Sundhed
Irriterende
Supplier:   MP Biomedicals
1,3-Propansulton, semi-solid
Catalog # ICNA0215639125
Supplier:  MP Biomedicals
CAS Number:  1120-71-4
1,3-Propansulton, semi-solid

Glass bottle for solids, 25 g

CAS Number:  1120-71-4
Price
707,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

1,3-Propane Sultone was used as quaternisation agent to transit the PDMAEMA into poly(zwitterions) through surface reaction by treating membranes under a mild condition.

1,3-Propane Sultone was used as quaternisation agent to transit the PDMAEMA into poly(zwitterions) through surface reaction by treating membranes under a mild condition.

Soluble in cold water, hot water; readily in ketones, esters and aromatic hydrocarbons; insoluble in aliphatic hydrocarbons.

Synonymer:  1,2-Oxathiolane 2,2-dioxide
  • CAS nummer:1120-71-4
  • Formel:C₃H₆O₃S
  • MDL nummer:MFCD00005355
  • Molecular Weight:122.14 g/mol
  • Kogende Pt:298 °C (1013 hPa)
  • Smeltende Pt:30…33 °C (lit.)
  • Tæthed:1,392 g/ml
  • Opbevaringstemperatur:Køleskab
  • EINECS:214-317-9
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer