Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Gentamicin sulfat

Glass bottle for solids, 1 g
Katalog # ICNA0219005701
Sundhed
Leverandør:   MP Biomedicals
Gentamicin sulfat
Katalog # ICNA0219005701
Leverandør:  MP Biomedicals
CAS nummer:  1405-41-0
Gentamicin sulfat

Glass bottle for solids, 1 g

CAS nummer:  1405-41-0
Price
1.320,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Gentamicin sulpahte is used as a selection agent (gentamicin-resistance gene) in molecular biology applications.It is used to treat many types of bacterial infections, particularly those caused by gram-negative organisms.

It causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site. The bactericidal effect of gentamicin on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilises the membrane, and forms holes in the cell surface. Antimicrobial spectrum: Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria.

H2O: soluble 50 mg/ml (As a stock solution. Stock solutions should be stored at −20 °C.) (Lit.)

Synonymer:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS nummer:1405-41-0
  • MDL nummer:MFCD00270181
  • Merck Index:13,04403
  • Smeltende Pt:218…237 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:215-778-9
Specifikationer

Specifikationstestresultater

  • Synonyms:
    Garamycin; Gentamicin sulphate salt; Gentiomycin C
  • Form:
    White powder
  • Storage:
    Store at +4 °C
  • Loss on Drying:
    ≤18,0 %
  • Optical Rotation:
    107° to +121° (10 mg/ml, water)
  • pH:
    3,5 - 5,5 (1 in 25, in water)
  • Potency:
    ≥590 µg/mg

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer