Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(R)-(+)-Limonen, liquid

(R)-(+)-Limonen, liquid

500 mL
Katalog # ICNA0215523490
Brandfarlig
Irriterende
Miljø
Leverandør:   MP Biomedicals
(R)-(+)-Limonen, liquid
Katalog # ICNA0215523490
Leverandør:  MP Biomedicals
CAS nummer:  5989-27-5
(R)-(+)-Limonen, liquid

500 mL

CAS nummer:  5989-27-5
Price
722,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

D-Limonene is a chiral intermediate for natural product synthesis. It is used as a renewably based solvent in cleaning products.

D-Limonene is a monoterpene found to prevent mammary cancer by inducing hepatic glutathione-S- transferase and uridine diphosphoglucuronosyl transferase. D-Limonene is an isoprenylation inhibitor of small G-proteins.

Limonene is a cyclie terpene from Chinese medicinal herb essential oils used in the synthesis of carvone. Limonene may be used as a shrinking agent to dissolve polystyrene. Limonene may be used in various insecticidal and insect repellant applications.

Miscible with alcohol, with most fixed oils, mineral oil; very slightly soluble in glycerin; practically insoluble in water and in propyiene glycol.

Synonymer:  (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexen, (R)-(+)-Dipenten, (R)-(+)-p-Mentha-1,8-dien, D(+)-Limonen
  • CAS nummer:5989-27-5
  • Formel:C₁₀H₁₆
  • MDL nummer:MFCD00062991
  • Molecular Weight:136.24 g/mol
  • Tæthed:0,84 g/ml
  • Opbevaringstemperatur:Stue temperatur
  • FN:2052
  • ADR:3,III

Documents

Specifikationer

Specifikationer

  • Pakn:
    500 mL

Specifikationer