Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(R)-(+)-Limonen, liquid

(R)-(+)-Limonen, liquid

100 mL
Catalog # ICNA0215523480
Flammable
Irritant
Environment
Supplier:   MP Biomedicals
(R)-(+)-Limonen, liquid
Catalog # ICNA0215523480
Supplier:  MP Biomedicals
CAS Number:  5989-27-5
(R)-(+)-Limonen, liquid

100 mL

CAS Number:  5989-27-5
Price
419,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

D-Limonene is a chiral intermediate for natural product synthesis. It is used as a renewably based solvent in cleaning products.

D-Limonene is a monoterpene found to prevent mammary cancer by inducing hepatic glutathione-S- transferase and uridine diphosphoglucuronosyl transferase. D-Limonene is an isoprenylation inhibitor of small G-proteins.

Limonene is a cyclie terpene from Chinese medicinal herb essential oils used in the synthesis of carvone. Limonene may be used as a shrinking agent to dissolve polystyrene. Limonene may be used in various insecticidal and insect repellant applications.

Miscible with alcohol, with most fixed oils, mineral oil; very slightly soluble in glycerin; practically insoluble in water and in propyiene glycol.

Synonyms:  (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexen, (R)-(+)-Dipenten, (R)-(+)-p-Mentha-1,8-dien, D(+)-Limonen
  • CAS Number:5989-27-5
  • Formula:C₁₀H₁₆
  • MDL Number:MFCD00062991
  • Molecular Weight:136.24 g/mol
  • Density:0,84 g/ml
  • Storage Temperature:Stue temperatur
  • UN:2052
  • ADR:3,III

Documents

Specifications

Specifications

  • Pakn:
    100 mL

Specifications