Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Oliesyre, liquid, Practical grade

Oliesyre, liquid, Practical grade

1 kg
Katalog # ICNA0210250091
Leverandør:   MP Biomedicals
Oliesyre, liquid, Practical grade
Katalog # ICNA0210250091
Leverandør:  MP Biomedicals
CAS nummer:  112-80-1
Oliesyre, liquid, Practical grade
Oliesyre, liquid, Practical grade

1 kg

CAS nummer:  112-80-1
Price
869,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Oleic acid is a mono-unsaturated omega-9 fatty acid created by dehydrogenation of stearic acid. It is then further dehydrogenated and elongated to create members of the n-9 family of fatty acids.

Oleic acid is used as emollient. Small amounts of oleic acid are used as an excipient in pharmaceuticals. It is used as an emulsifying or solubilizing agent in aerosol products.

Oleic Acid activates protein kinase C in hepatocytes. It uncouples oxidative phosphorylati It inhibits 2,4-dinitrophenol -stimulated ATPase. Action reversed by adding serum albumin.

Soluble in methanol (40 mg/ml - clear solution), chloroform, benzene or fixed and volatile oils. Insoluble in water.

Advarsel: Research Use Only (RUO).

Synonymer:  (9Z)-9-Octadecenoic acid
  • CAS nummer:112-80-1
  • Formel:C₁₈H₃₄O₂
  • MDL nummer:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.