Docusate natrium, white waxy solid
500 g
Katalog # ICNA0210157590
Leverandør: MP Biomedicals
Docusate natrium, white waxy solid
Katalog # ICNA0210157590
Leverandør: MP Biomedicals
CAS nummer: 577-11-7
Gå til:
Product Details & Documents
Dioctyl sulphosuccinate sodium salt is used to make a microemulsion with CAPSO for the electrophoresis detection of natural and synthetic estrogens. Used to prepare reverse micelles.
Dioctyl sodium sulphosuccinate (DSS) dissolves slowly in water; at 25 °C to the extent of 1,5 gm/100 cc; at 70 °C, 5,5 gm/100 cc. It dissolves in oils, hydrocarbons, fats and waxs by heating above 75 °C and remains in solution when cooled to room temperature. At room temperature, it is readily soluble in most organic solvents, both polar and non-polar. Soluble in carbon tetrachloride, petroleum ether, naphtha, xylene, dibutyl phthalate, liquid petroleum, acetone, alcohol, vegetable oils.
Advarsel: Research Use Only (RUO).
Synonymer: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
- CAS nummer:577-11-7
- Formel:C₂₀H₃₇NaO₇S
- MDL nummer:MFCD00012455
- Molecular Weight:444.57 g/mol
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationstestresultater
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH (1% aq soln):5 - 8
Specifikationer
- Pakn:500 g



