Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(S)-(-)-Æblesyre ≥99%, powder

Glass bottle for solids, 100 g
Katalog # ICNA0210223780
Irriterende
Leverandør:   MP Biomedicals
(S)-(-)-Æblesyre ≥99%, powder
Katalog # ICNA0210223780
Leverandør:  MP Biomedicals
CAS nummer:  97-67-6
(S)-(-)-Æblesyre ≥99%, powder

Glass bottle for solids, 100 g

CAS nummer:  97-67-6
Price
613,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.

L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.

Soluble in 100 mg/ml - clear to very slightly hazy, colourless.

Synonymer:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS nummer:97-67-6
  • Formel:C₄H₆O₅
  • MDL nummer:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:13,05730
  • Kogende Pt:322 °C (1013 hPa)
  • Smeltende Pt:101…104 °C
  • Tæthed:1,595 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-601-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer