(S)-(-)-Æblesyre ≥99%, powder
Glass bottle for solids, 25 g
Katalog # ICNA0210223725
Leverandør: MP Biomedicals
(S)-(-)-Æblesyre ≥99%, powder
Katalog # ICNA0210223725
Leverandør: MP Biomedicals
CAS nummer: 97-67-6
Gå til:
Product Details & Documents
L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.
L-Malic Acid is a dicarboxylic acid that is made by all living organisms. The malate anion is an intermediate in the citric acid cycle.
Soluble in 100 mg/ml - clear to very slightly hazy, colourless.
Synonymer: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
- CAS nummer:97-67-6
- Formel:C₄H₆O₅
- MDL nummer:MFCD00064213
- Molecular Weight:134.09 g/mol
- Merck Index:13,05730
- Kogende Pt:322 °C (1013 hPa)
- Smeltende Pt:101…104 °C
- Tæthed:1,595 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-601-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



