Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Polymyxin Bsulfat

Polymyxin Bsulfat

25 Mu
Katalog # ICNA0210056525
Leverandør:   MP Biomedicals
Polymyxin Bsulfat
Katalog # ICNA0210056525
Leverandør:  MP Biomedicals
CAS nummer:  1405-20-5
Polymyxin Bsulfat

25 Mu

CAS nummer:  1405-20-5
Price
3.890,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Polymyxin B sulphate is a mixture of Polymyxin B1 and Polymyxin B2 sulphate, obtained from Bacillus polymyxia strains. Polymyxin B inhibits gram-negative organisms in vitro wherein it binds to and interferes with the permeability of the bacterial cytoplasmic membrane and is also reported to inhibit Ca2+ dependent protein kinase C (PKC). Effective concentration is typically 50 mg/l.

  • CAS nummer:1405-20-5
  • Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  • MDL nummer:MFCD00131991
  • Molecular Weight:1385.61 g/mol
Specifikationer

Specifikationer

  • Pakn:
    25 Mu

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.