Orders LinkContactUsLinkComponent
Polymyxin Bsulfat
5 Mu
  ICNA0210056505
Polymyxin Bsulfat
  ICNA0210056505
 :  1405-20-5
Polymyxin Bsulfat
5 Mu
 :  1405-20-5
1.350,00 kr
1 * 1 Each
Polymyxin B sulphate is a mixture of Polymyxin B1 and Polymyxin B2 sulphate, obtained from Bacillus polymyxia strains. Polymyxin B inhibits gram-negative organisms in vitro wherein it binds to and interferes with the permeability of the bacterial cytoplasmic membrane and is also reported to inhibit Ca2+ dependent protein kinase C (PKC). Effective concentration is typically 50 mg/l.
  •  :1405-20-5
  •  :C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  •  :MFCD00131991
  •  :1385.61 g/mol
  • Pakn:
    5 Mu