Polymyxin Bsulfat
5 Mu
ICNA0210056505
Polymyxin B sulphate is a mixture of Polymyxin B1 and Polymyxin B2 sulphate, obtained from Bacillus polymyxia strains. Polymyxin B inhibits gram-negative organisms in vitro wherein it binds to and interferes with the permeability of the bacterial cytoplasmic membrane and is also reported to inhibit Ca2+ dependent protein kinase C (PKC). Effective concentration is typically 50 mg/l.
- :1405-20-5
- :C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
- :MFCD00131991
- :1385.61 g/mol
- Pakn:5 Mu