Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Polymyxin Bsulfat

1 Mu
Katalog # ICNA0210056501
Leverandør:   MP Biomedicals
Polymyxin Bsulfat
Katalog # ICNA0210056501
Leverandør:  MP Biomedicals
CAS nummer:  1405-20-5
Polymyxin Bsulfat

1 Mu

CAS nummer:  1405-20-5
Price
388,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Polymyxin B sulphate is a mixture of Polymyxin B1 and Polymyxin B2 sulphate, obtained from Bacillus polymyxia strains. Polymyxin B inhibits gram-negative organisms in vitro wherein it binds to and interferes with the permeability of the bacterial cytoplasmic membrane and is also reported to inhibit Ca2+ dependent protein kinase C (PKC). Effective concentration is typically 50 mg/l.

  • CAS nummer:1405-20-5
  • Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  • MDL nummer:MFCD00131991
  • Molecular Weight:1385.61 g/mol
Specifikationer

Specifikationer

  • Pakn:
    1 Mu

Specifikationer