n-Hexadecan ≥99%, liquid
1 L
Katalog # ICNA0219521891
Leverandør: MP Biomedicals
Gå til:
Product Details & Documents
Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.
Soluble in ether; insoluble in water.
Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)
Synonymer: Hexadecane, Cetane
- CAS nummer:544-76-3
- Formel:H₃C(CH₂)₁₄CH₃
- MDL nummer:MFCD00008998
- Molecular Weight:226.45 g/mol
Specifikationer
Specifikationer
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



