Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexadecan ≥99%, liquid

n-Hexadecan ≥99%, liquid

500 mL
Katalog # ICNA0219521890
Leverandør:   MP Biomedicals
n-Hexadecan ≥99%, liquid
Katalog # ICNA0219521890
Leverandør:  MP Biomedicals
CAS nummer:  544-76-3
n-Hexadecan ≥99%, liquid
n-Hexadecan ≥99%, liquid

500 mL

CAS nummer:  544-76-3
Price
1.180,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.

Soluble in ether; insoluble in water.

Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)

Synonymer:  Hexadecane, Cetane
  • CAS nummer:544-76-3
  • Formel:H₃C(CH₂)₁₄CH₃
  • MDL nummer:MFCD00008998
  • Molecular Weight:226.45 g/mol
Specifikationer

Specifikationer

  • Pakn:
    500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.