n-Hexadecan ≥99%, liquid
100 mL
Catalog # ICNA0219521880
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP.
Soluble in ether; insoluble in water.
Transition temp.- solidification point 17.5 - 18.5 °C (Lit.)
Synonyms: Hexadecane, Cetane
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
Specifications
Specifications
- Pakn:100 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



