Orders LinkContactUsLinkComponent
Accessories for refrigerated circulating bath VersaCool™
Accessories for refrigerated circulating bath VersaCool™
Stainless steel, adjustable VersaRack, holds up to 75 test tubes of 10 mm diameter, 48 test tubes of 16 mm diameter and 21 test tubes of 25 mm diameter
Katalog # HAKA230000001
Leverandør:   THERMO ELECTRON LED
Accessories for refrigerated circulating bath VersaCool™
Katalog # HAKA230000001
Leverandør:  THERMO ELECTRON LED
Accessories for refrigerated circulating bath VersaCool™
Stainless steel, adjustable VersaRack, holds up to 75 test tubes of 10 mm diameter, 48 test tubes of 16 mm diameter and 21 test tubes of 25 mm diameter
Price
7.460,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Stainless steel, adjustable VersaRack, holds up to 75 test tubes of 10 mm diameter, 48 test tubes of 16 mm diameter and 21 test tubes of 25 mm diameter
  • Seq:
    07