Cleaning agents, elma tec clean
elma tec clean A2, for cleaning non ferrous metals, precious metals, brass, nickel, titanium; check zinc and aluminium for resistance before use!, 11,0
Powerful cleaners for ultrasonic baths suitable for many industrial cleaning tasks
- prolongs the service life of the ultrasonic unit
- Highly efficient concentrate
- Long shelf life
- Beskrivelse:elma tec clean A2, for cleaning non ferrous metals, precious metals, brass, nickel, titanium; check zinc and aluminium for resistance before use!
- pH-værdi:11,0