Orders LinkContactUsLinkComponent
Cleaning agents, elma tec clean
elma tec clean A2, for cleaning non ferrous metals, precious metals, brass, nickel, titanium; check zinc and aluminium for resistance before use!, 11,0
  ELMA8000112
Cleaning agents, elma tec clean
  ELMA8000112
Cleaning agents, elma tec clean
Cleaning agents, elma tec clean
elma tec clean A2, for cleaning non ferrous metals, precious metals, brass, nickel, titanium; check zinc and aluminium for resistance before use!, 11,0
418,00 kr
1 * 2,5 L
Powerful cleaners for ultrasonic baths suitable for many industrial cleaning tasks
  • prolongs the service life of the ultrasonic unit
  • Highly efficient concentrate
  • Long shelf life
  • Beskrivelse:
    elma tec clean A2, for cleaning non ferrous metals, precious metals, brass, nickel, titanium; check zinc and aluminium for resistance before use!
  • pH-værdi:
    11,0
Accessories
  •  
    Beskrivelse
  • ELMA8000003927
    Hane til 5 l og 10 l beholdere
    1 * 1 Items - 117,00 kr
  •  
    Beskrivelse
  • ELMA8000003928
    Hane til 25 l beholdere
    1 * 1 Items - 117,00 kr