Volumetric flasks, class A, white markings
1000 ml, 24/29, 0,40 ± ml
Katalog # POULRB258/AM/WAC/K
Leverandør: Poulten & Graf
Gå til:
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards
Specifikationer
- NS:24/29
- Kapacitet:1000 ml
- Seq:23
- Tolerance:0,40 ± ml