Volumetric flasks, class A, white markings
250 ml, 14/23, 0,15 ± ml
Katalog # POULRB258/AM/WAC/I
Leverandør: Poulten & Graf
Gå til:
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards
Specifikationer
- NS:14/23
- Kapacitet:250 ml
- Seq:21
- Tolerance:0,15 ± ml