Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
200 ml, 14/23, 0,15 ± ml
Katalog # POULRB258/AM/WAC/H
Leverandør:   Poulten & Graf
Volumetric flasks, class A, white markings
Katalog # POULRB258/AM/WAC/H
Leverandør:  Poulten & Graf
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
200 ml, 14/23, 0,15 ± ml
Price
785,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
  • Easy to read white markings
  • Very good chemical resistance
  • Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards
Specifikationer
  • NS:
    14/23
  • Kapacitet:
    200 ml
  • Seq:
    20
  • Tolerance:
    0,15 ± ml