Volumetric flasks, class A, white markings
50 ml, 12/21, 0,04 ± ml
Katalog # POULRB258/AM/WAC/F
Leverandør: Poulten & Graf
Gå til:
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
: Acc. to ISO and ASTM or USP standards
Specifikationer
- NS:12/21
- Kapacitet:50 ml
- Seq:18
- Tolerance:0,04 ± ml