Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
20 ml, 10/19, 0,04 ± ml
Catalog # POULRB258/AM/WAC/D
Supplier:   Poulten & Graf
Volumetric flasks, class A, white markings
Catalog # POULRB258/AM/WAC/D
Supplier:  Poulten & Graf
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
20 ml, 10/19, 0,04 ± ml
Price
1.850,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
  • Easy to read white markings
  • Very good chemical resistance
  • Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards
Specifications
  • NS:
    10/19
  • Kapacitet:
    20 ml
  • Seq:
    16
  • Tolerance:
    0,04 ± ml