Volumetric flasks, class A, white markings
20 ml, 10/19, 0,04 ± ml
Catalog # POULRB258/AM/WAC/D
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards
Specifications
- NS:10/19
- Kapacitet:20 ml
- Seq:16
- Tolerance:0,04 ± ml