Cables
InLab® cable S7-DIN, 3 m
Katalog # 661-0262
Leverandør: Mettler - Toledo
Gå til:
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Mettler InLab® Sensors
Produktlitteratur
Specifikationer
- Beskrivelse:InLab® cable S7-DIN
- Længde:3 m
- Seq:163