Orders LinkContactUsLinkComponent
[EN]CRATE STACKABLE W/LID 20L 38X28.5X29 1 * 1 Items
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
[EN]CRATE STACKABLE W/LID 20L 38X28.5X29 1 * 1 Items
Catalog # LAMA1908
Supplier:   LAMAPLAST
[EN]CRATE STACKABLE W/LID 20L 38X28.5X29 1 * 1 Items
Catalog # LAMA1908
Supplier:  LAMAPLAST
[EN]CRATE STACKABLE W/LID 20L 38X28.5X29 1 * 1 Items
Price
467,00 kr