Cables
InLab® cable MultiPin-DIN/4mm, 1,2 m
Katalog # 661-0254
Leverandør: Mettler - Toledo
Gå til:
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Mettler InLab® Sensors
Produktlitteratur
Specifikationer
- Beskrivelse:InLab® cable MultiPin-DIN/4mm
- Længde:1,2 m
- Seq:155